Assay Details
| Gene Symbol: SCRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332054.1 | NP_001320452.1 | 2 - 3 | 1792 | 70 | |
| NM_101157.3 | NP_172746.2 | 3 - 4 | 1725 | 70 | ||
| GenBank mRNA | AK118011.1 | - | 3 - 4 | 1342 | 70 | |
| BT005377.1 | - | 3 - 4 | 1271 | 70 | ||
| EU369670.1 | - | 3 - 4 | 1406 | 70 | ||
More Information
Set Membership: |
|

SCRM2
Genomic Map