Assay Details
| Gene Symbol: AT1G12930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101164.5 | NP_563920.1 | 19 - 20 | 2381 | 148 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G12930| Gene Symbol: AT1G12930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101164.5 | NP_563920.1 | 19 - 20 | 2381 | 148 | |
Set Membership: |
|