Assay Details
| Gene Symbol: RSH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101166.4 | NP_172755.1 | 6 - 7 | 1545 | 112 | |
| GenBank mRNA | BT023437.1 | - | 6 - 7 | 1416 | 112 | |
More Information
Set Membership: |
|

RSH2
Genomic Map