Assay Details
| Gene Symbol: AT1G13160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101186.5 | NP_172775.1 | 8 - 9 | 1260 | 66 | |
| GenBank mRNA | AK226773.1 | - | 8 - 9 | 1262 | 66 | |
More Information
Set Membership: |
|

AT1G13160
Genomic Map