Assay Details
| Gene Symbol: HRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101201.2 | NP_563926.1 | 2 - 3 | 524 | 91 | |
| GenBank mRNA | AF367336.1 | - | 2 - 3 | 512 | 91 | |
| AY124865.1 | - | 2 - 3 | 425 | 91 | ||
More Information
Set Membership: |
|

HRS1
Genomic Map