Assay Details
| Gene Symbol: AHP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101204.3 | NP_172791.1 | 5 - 6 | 651 | 121 | |
| GenBank mRNA | AJ627484.1 | - | 5 - 6 | 578 | 121 | |
| AK176610.1 | - | 5 - 6 | 651 | 121 | ||
| AK227903.1 | - | 5 - 6 | 651 | 121 | ||
| AY225519.1 | - | 5 - 6 | 578 | 121 | ||
| BT025172.1 | - | 5 - 6 | 578 | 121 | ||
More Information
Set Membership: |
|

AHP2
Genomic Map