Assay Details
| Gene Symbol: ELF5A-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101261.3 | NP_172848.1 | 3 - 4 | 496 | 97 | |
| GenBank mRNA | AF296082.1 | - | 3 - 4 | 354 | 97 | |
| AY063780.1 | - | 3 - 4 | 339 | 97 | ||
| AY117272.1 | - | 3 - 4 | 299 | 97 | ||
More Information
Set Membership: |
|

ELF5A-1
Genomic Map