Assay Details
| Gene Symbol: AT1G14230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101289.5 | NP_563942.1 | 3 - 4 | 1155 | 86 | |
| GenBank mRNA | AY150499.1 | - | 3 - 4 | 826 | 86 | |
| JF830009.1 | - | 3 - 4 | 826 | 86 | ||
More Information
Set Membership: |
|

AT1G14230
Genomic Map