Assay Details
| Gene Symbol: AT1G14230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101289.5 | NP_563942.1 | 4 - 5 | 1202 | 140 | |
| GenBank mRNA | AY150499.1 | - | 4 - 5 | 873 | 140 | |
| JF830009.1 | - | 4 - 5 | 873 | 140 | ||
More Information
Set Membership: |
|

AT1G14230
Genomic Map