Assay Details
| Gene Symbol: AT1G14250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101291.3 | NP_172877.1 | 4 - 5 | 1001 | 113 | |
| GenBank mRNA | AK175135.1 | - | 4 - 5 | 895 | 113 | |
| AK175519.1 | - | 4 - 5 | 895 | 113 | ||
| BT010550.1 | - | 3 - 4 | 801 | 113 | ||
| JF830010.1 | - | 3 - 4 | 801 | 113 | ||
More Information
Set Membership: |
|

AT1G14250
Genomic Map