Assay Details
| Gene Symbol: APK2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101304.4 | NP_172889.1 | 1 - 2 | 438 | 71 | |
| GenBank mRNA | AF334731.1 | - | 1 - 2 | 70 | 71 | |
| AY059894.1 | - | 1 - 2 | 262 | 71 | ||
| D88206.1 | - | 1 - 2 | 246 | 71 | ||
More Information
Set Membership: |
|

APK2A
Genomic Map