Assay Details
| Gene Symbol: APK2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101304.4 | NP_172889.1 | 4 - 5 | 1056 | 112 | |
| GenBank mRNA | AF334731.1 | - | 4 - 5 | 688 | 112 | |
| AY059894.1 | - | 4 - 5 | 880 | 112 | ||
| D88206.1 | - | 4 - 5 | 864 | 112 | ||
More Information
Set Membership: |
|

APK2A
Genomic Map