Assay Details
| Gene Symbol: MIOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160865.1 | NP_001154337.1 | 6 - 7 | 798 | 101 | |
| NM_001332122.1 | NP_001320938.1 | 6 - 7 | 1108 | 101 | ||
| NM_001332123.1 | NP_001320937.1 | 6 - 7 | 1108 | 101 | ||
| NM_101319.4 | NP_172904.2 | 7 - 8 | 1008 | 101 | ||
| GenBank mRNA | AK175115.1 | - | 6 - 7 | 870 | 101 | |
| AK176690.1 | - | 6 - 7 | 870 | 101 | ||
| AY136388.1 | - | 6 - 7 | 811 | 101 | ||
| BT000187.1 | - | 6 - 7 | 751 | 101 | ||
More Information
Set Membership: |
|

MIOX1
Genomic Map