Assay Details
| Gene Symbol: DL1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101352.3 | NP_172936.1 | 13 - 14 | 1610 | 67 | |
| GenBank mRNA | AJ304841.1 | - | 13 - 14 | 1534 | 67 | |
| AY039955.1 | - | 13 - 14 | 1495 | 67 | ||
| AY150494.1 | - | 13 - 14 | 1334 | 67 | ||
More Information
Set Membership: |
|

DL1C
Genomic Map