Assay Details
| Gene Symbol: DL1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101352.3 | NP_172936.1 | 15 - 16 | 1995 | 75 | |
| GenBank mRNA | AJ304841.1 | - | 15 - 16 | 1919 | 75 | |
| AY039955.1 | - | 15 - 16 | 1880 | 75 | ||
| AY150494.1 | - | 15 - 16 | 1719 | 75 | ||
More Information
Set Membership: |
|

DL1C
Genomic Map