Assay Details
| Gene Symbol: AT1G21660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102015.3 | NP_173585.1 | 1 - 2 | 885 | 80 | |
| GenBank mRNA | AY099577.1 | - | 1 - 2 | 806 | 80 | |
| BT010382.1 | - | 1 - 2 | 581 | 80 | ||
More Information
Set Membership: |
|

AT1G21660
Genomic Map