Assay Details
| Gene Symbol: AT1G21660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102015.3 | NP_173585.1 | 3 - 4 | 1287 | 108 | |
| GenBank mRNA | AY099577.1 | - | 3 - 4 | 1208 | 108 | |
| BT010382.1 | - | 3 - 4 | 983 | 108 | ||
More Information
Set Membership: |
|

AT1G21660
Genomic Map