Assay Details
| Gene Symbol: AT1G21660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102015.3 | NP_173585.1 | 4 - 5 | 1404 | 80 | |
| GenBank mRNA | AY099577.1 | - | 4 - 5 | 1325 | 80 | |
| BT010382.1 | - | 4 - 5 | 1100 | 80 | ||
More Information
Set Membership: |
|

AT1G21660
Genomic Map