Assay Details
| Gene Symbol: AT1G21730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102022.4 | NP_173592.3 | 21 - 22 | 2476 | 121 | |
| GenBank mRNA | AB062738.1 | - | 21 - 22 | 2476 | 121 | |
More Information
Set Membership: |
|

AT1G21730
Genomic Map