Assay Details
| Gene Symbol: AT1G21790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102028.3 | NP_564152.1 | 2 - 3 | 806 | 101 | |
| GenBank mRNA | AY063913.1 | - | 2 - 3 | 640 | 101 | |
| AY087660.1 | - | 2 - 3 | 710 | 101 | ||
| AY091268.1 | - | 2 - 3 | 537 | 101 | ||
More Information
Set Membership: |
|

AT1G21790
Genomic Map