Assay Details
| Gene Symbol: BS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106110.3 | NP_177590.1 | 1 - 2 | 287 | 72 | |
| GenBank mRNA | AK227395.1 | - | 1 - 2 | 287 | 72 | |
| AY088286.1 | - | 1 - 2 | 287 | 72 | ||
| BT003046.1 | - | 1 - 2 | 134 | 72 | ||
More Information
Set Membership: |
|

BS1
Genomic Map