Assay Details
| Gene Symbol: MYB31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334659.1 | NP_001320340.1 | 1 - 2 | 687 | 72 | |
| NM_106123.4 | NP_177603.1 | 2 - 3 | 611 | 72 | ||
| GenBank mRNA | BT029481.1 | - | 2 - 3 | 264 | 72 | |
More Information
Set Membership: |
|

MYB31
Genomic Map