Assay Details
| Gene Symbol: TRZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106128.3 | NP_177608.2 | 4 - 5 | 834 | 153 | |
| GenBank mRNA | AJ428989.1 | - | 4 - 5 | 503 | 153 | |
| BT025035.1 | - | 4 - 5 | 503 | 153 | ||
More Information
Set Membership: |
|

TRZ1
Genomic Map