Assay Details
| Gene Symbol: BOR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334667.1 | NP_001319383.1 | 13 - 14 | 2124 | 72 | |
| NM_001334668.1 | NP_001322361.1 | 10 - 11 | 2798 | 72 | ||
| NM_001334669.1 | NP_001322362.1 | 11 - 12 | 2714 | 72 | ||
| NM_001334670.1 | NP_001322363.1 | 12 - 13 | 2566 | 72 | ||
| NM_001334671.1 | NP_001322364.1 | 12 - 13 | 2255 | 72 | ||
| NM_001334672.1 | NP_001322360.1 | 13 - 14 | 2136 | 72 | ||
| NM_106139.3 | NP_177619.2 | 12 - 13 | 2642 | 72 | ||
More Information
Set Membership: |
|

BOR5
Genomic Map