Assay Details
| Gene Symbol: BOR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334667.1 | NP_001319383.1 | 10 - 11 | 1387 | 114 | |
| NM_001334668.1 | NP_001322361.1 | 7 - 8 | 2061 | 114 | ||
| NM_001334669.1 | NP_001322362.1 | 8 - 9 | 1977 | 114 | ||
| NM_001334670.1 | NP_001322363.1 | 9 - 10 | 1829 | 114 | ||
| NM_001334671.1 | NP_001322364.1 | 9 - 10 | 1518 | 114 | ||
| NM_001334672.1 | NP_001322360.1 | 10 - 11 | 1399 | 114 | ||
| NM_106139.3 | NP_177619.2 | 9 - 10 | 1905 | 114 | ||
More Information
Set Membership: |
|

BOR5
Genomic Map