Assay Details
| Gene Symbol: BBX21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106207.4 | NP_177686.1 | 1 - 2 | 421 | 137 | |
| GenBank mRNA | AB493536.1 | - | 1 - 2 | 186 | 137 | |
More Information
Set Membership: |
|

BBX21
Genomic Map