Assay Details
| Gene Symbol: XRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106217.3 | NP_565114.1 | 13 - 14 | 1836 | 114 | |
| GenBank mRNA | AF286719.1 | - | 13 - 14 | 1755 | 114 | |
More Information
Set Membership: |
|

XRN3
Genomic Map