Assay Details
| Gene Symbol: XRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106217.3 | NP_565114.1 | 3 - 4 | 540 | 83 | |
| GenBank mRNA | AF286719.1 | - | 3 - 4 | 459 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

XRN3| Gene Symbol: XRN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106217.3 | NP_565114.1 | 3 - 4 | 540 | 83 | |
| GenBank mRNA | AF286719.1 | - | 3 - 4 | 459 | 83 | |
Set Membership: |
|