Assay Details
| Gene Symbol: VPS35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334699.1 | NP_001321274.1 | 11 - 12 | 1134 | 73 | |
| NM_106235.5 | NP_177713.3 | 16 - 17 | 1759 | 73 | ||
More Information
Set Membership: |
|

VPS35B
Genomic Map