Assay Details
| Gene Symbol: VPS35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106235.5 | NP_177713.3 | 1 - 2 | 171 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VPS35B| Gene Symbol: VPS35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106235.5 | NP_177713.3 | 1 - 2 | 171 | 84 | |
Set Membership: |
|