Assay Details
| Gene Symbol: VPS35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334699.1 | NP_001321274.1 | 2 - 3 | 178 | 134 | |
| NM_106235.5 | NP_177713.3 | 7 - 8 | 803 | 134 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001334699.1 transcripts.
Set Membership: |
|

VPS35B
Genomic Map