Assay Details
| Gene Symbol: VPS35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334699.1 | NP_001321274.1 | 4 - 5 | 388 | 118 | |
| NM_106235.5 | NP_177713.3 | 9 - 10 | 1013 | 118 | ||
More Information
Set Membership: |
|

VPS35B
Genomic Map