Assay Details
| Gene Symbol: VPS35B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334699.1 | NP_001321274.1 | 5 - 6 | 440 | 106 | |
| NM_106235.5 | NP_177713.3 | 10 - 11 | 1065 | 106 | ||
More Information
Set Membership: |
|

VPS35B
Genomic Map