Assay Details
| Gene Symbol: ATA27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106244.3 | NP_177722.1 | 11 - 12 | 1330 | 65 | |
| GenBank mRNA | BT002735.1 | - | 11 - 12 | 1274 | 65 | |
More Information
Set Membership: |
|

ATA27
Genomic Map