Assay Details
| Gene Symbol: ATA27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106244.3 | NP_177722.1 | 12 - 13 | 1448 | 108 | |
| GenBank mRNA | BT002735.1 | - | 12 - 13 | 1392 | 108 | |
More Information
Set Membership: |
|

ATA27
Genomic Map