Assay Details
| Gene Symbol: ATA27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106244.3 | NP_177722.1 | 2 - 3 | 290 | 64 | |
| GenBank mRNA | BT002735.1 | - | 2 - 3 | 234 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATA27| Gene Symbol: ATA27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106244.3 | NP_177722.1 | 2 - 3 | 290 | 64 | |
| GenBank mRNA | BT002735.1 | - | 2 - 3 | 234 | 64 | |
Set Membership: |
|