Assay Details
| Gene Symbol: SKP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106245.5 | NP_565123.1 | 1 - 2 | 512 | 110 | |
| GenBank mRNA | AF059294.1 | - | 1 - 2 | 381 | 110 | |
| AY080884.1 | - | 1 - 2 | 376 | 110 | ||
| AY113971.1 | - | 1 - 2 | 316 | 110 | ||
| U60981.1 | - | 1 - 2 | 365 | 110 | ||
| U97020.1 | - | 1 - 2 | 385 | 110 | ||
More Information
Set Membership: |
|

SKP1
Genomic Map