Assay Details
| Gene Symbol: AMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334710.1 | NP_001323338.1 | 10 - 11 | 1370 | 68 | |
| NM_106262.4 | NP_177740.1 | 10 - 11 | 1005 | 68 | ||
| GenBank mRNA | AK221564.1 | - | 10 - 11 | 1005 | 68 | |
| AY084871.1 | - | 10 - 11 | 1001 | 68 | ||
| BT025560.1 | - | 9 - 10 | 918 | 68 | ||
More Information
Set Membership: |
|

AMY2
Genomic Map