Assay Details
| Gene Symbol: AMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334710.1 | NP_001323338.1 | 12 - 13 | 1496 | 96 | |
| NM_106262.4 | NP_177740.1 | 12 - 13 | 1131 | 96 | ||
| GenBank mRNA | AK221564.1 | - | 12 - 13 | 1131 | 96 | |
| AY084871.1 | - | 12 - 13 | 1127 | 96 | ||
| BT025560.1 | - | 11 - 12 | 1044 | 96 | ||
More Information
Set Membership: |
|

AMY2
Genomic Map