Assay Details
| Gene Symbol: AMY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334710.1 | NP_001323338.1 | 8 - 9 | 1162 | 77 | |
| NM_106262.4 | NP_177740.1 | 8 - 9 | 797 | 77 | ||
| GenBank mRNA | AK221564.1 | - | 8 - 9 | 797 | 77 | |
| AY084871.1 | - | 8 - 9 | 793 | 77 | ||
| BT025560.1 | - | 7 - 8 | 710 | 77 | ||
More Information
Set Membership: |
|

AMY2
Genomic Map