Assay Details
| Gene Symbol: ECH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106264.3 | NP_177742.2 | 2 - 3 | 138 | 82 | |
| GenBank mRNA | AY070763.1 | - | 2 - 3 | 114 | 82 | |
| AY093712.1 | - | 2 - 3 | 75 | 82 | ||
More Information
Set Membership: |
|

ECH2
Genomic Map