Assay Details
| Gene Symbol: ECH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106264.3 | NP_177742.2 | 8 - 9 | 658 | 81 | |
| GenBank mRNA | AY070763.1 | - | 8 - 9 | 634 | 81 | |
| AY093712.1 | - | 8 - 9 | 595 | 81 | ||
More Information
Set Membership: |
|

ECH2
Genomic Map