Assay Details
| Gene Symbol: ECH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106264.3 | NP_177742.2 | 9 - 10 | 730 | 126 | |
| GenBank mRNA | AY070763.1 | - | 9 - 10 | 706 | 126 | |
| AY093712.1 | - | 9 - 10 | 667 | 126 | ||
More Information
Set Membership: |
|

ECH2
Genomic Map