Assay Details
| Gene Symbol: DWA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106276.5 | NP_177753.1 | 8 - 9 | 932 | 81 | |
| GenBank mRNA | AK175439.1 | - | 8 - 9 | 773 | 81 | |
| AK176650.1 | - | 8 - 9 | 773 | 81 | ||
| BT010880.1 | - | 8 - 9 | 762 | 81 | ||
More Information
Set Membership: |
|

DWA2
Genomic Map