Assay Details
| Gene Symbol: PHT1;9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106293.4 | NP_177769.1 | 1 - 2 | 899 | 63 | |
| GenBank mRNA | AK228274.1 | - | 1 - 2 | 832 | 63 | |
| BT008324.1 | - | 1 - 2 | 711 | 63 | ||
More Information
Set Membership: |
|

PHT1;9
Genomic Map