Assay Details
| Gene Symbol: HMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106299.4 | NP_177775.2 | 3 - 4 | 1705 | 85 | |
| GenBank mRNA | AY488113.1 | - | 3 - 4 | 1705 | 85 | |
More Information
Set Membership: |
|

HMG1
Genomic Map