Assay Details
| Gene Symbol: AT1G76670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334739.1 | NP_001319390.1 | 3 - 4 | 872 | 88 | |
| NM_106317.3 | NP_565138.1 | 3 - 4 | 940 | 88 | ||
| GenBank mRNA | AJ633721.1 | - | 3 - 4 | 545 | 88 | |
| AY039973.1 | - | 3 - 4 | 809 | 88 | ||
| AY058088.1 | - | 3 - 4 | 853 | 88 | ||
| AY074532.1 | - | 3 - 4 | 520 | 88 | ||
| BT000556.1 | - | 3 - 4 | 520 | 88 | ||
| KJ667158.1 | - | 3 - 4 | 520 | 88 | ||
| KP872769.1 | - | 3 - 4 | 520 | 88 | ||
More Information
Set Membership: |
|

AT1G76670
Genomic Map