Assay Details
| Gene Symbol: COG0212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106323.4 | NP_565139.1 | 5 - 6 | 1068 | 71 | |
| GenBank mRNA | BT024883.1 | - | 5 - 6 | 913 | 71 | |
More Information
Set Membership: |
|

COG0212
Genomic Map