Assay Details
| Gene Symbol: EC1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106325.2 | NP_177801.1 | 1 | 484 | 108 | |
| GenBank mRNA | AY735569.1 | - | 1 | 484 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EC1.1| Gene Symbol: EC1.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106325.2 | NP_177801.1 | 1 | 484 | 108 | |
| GenBank mRNA | AY735569.1 | - | 1 | 484 | 108 | |
Set Membership: |
|