Assay Details
| Gene Symbol: TY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106326.8 | NP_177802.2 | 1 - 2 | 399 | 110 | |
| GenBank mRNA | AK227422.1 | - | 1 - 2 | 305 | 110 | |
| BT010677.1 | - | 1 - 2 | 274 | 110 | ||
| BT010965.1 | - | 1 - 2 | 183 | 110 | ||
More Information
Set Membership: |
|

TY1
Genomic Map